Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145884.1 | NP_001139356.1 | 4 - 5 | 910 | 75 | |
NM_010580.2 | NP_034710.2 | 4 - 5 | 910 | 75 | ||
GenBank mRNA | AF022110.1 | - | 4 - 5 | 909 | 75 | |
AF043256.1 | - | 4 - 5 | 815 | 75 | ||
AF043257.1 | - | 4 - 5 | 815 | 75 | ||
AK087476.1 | - | 4 - 5 | 656 | 75 | ||
AK140251.1 | - | 4 - 5 | 822 | 75 | ||
AK143532.1 | - | 4 - 5 | 845 | 75 | ||
BC046280.1 | - | 4 - 5 | 819 | 6394641 | 75 | |
BC058246.1 | - | 4 - 5 | 874 | 6313668 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |