Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600005285.mm.1 |
NM_001205286.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005285.mm.1 |
NM_001205286.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005285.mm.1 |
NM_001205287.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005285.mm.1 |
NM_001205287.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005285.mm.1 |
NM_026407.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005285.mm.1 |
NM_026407.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005285.mm.1 |
TC1600000558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600005289.mm.1 |
NM_001205286.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005289.mm.1 |
NM_001205286.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005289.mm.1 |
NM_001205287.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005289.mm.1 |
NM_001205287.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005289.mm.1 |
NM_026407.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005289.mm.1 |
NM_026407.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005289.mm.1 |
TC1600000558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600005290.mm.1 |
NM_001205286.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005290.mm.1 |
NM_001205286.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005290.mm.1 |
NM_001205287.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005290.mm.1 |
NM_001205287.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005290.mm.1 |
NM_026407.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005290.mm.1 |
NM_026407.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600005290.mm.1 |
TC1600000558.mm.1 |
MTA TC |