Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028060.3 | NP_082336.3 | 7 - 8 | 1190 | 91 | |
GenBank mRNA | AK005182.1 | - | 7 - 8 | 986 | 91 | |
BC019202.1 | - | 7 - 8 | 992 | 92 | ||
BC050202.1 | - | 1 - 2 | 315 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |