Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177326.3 | NP_796300.1 | 13 - 14 | 1624 | 137 | |
GenBank mRNA | AK037275.1 | - | 13 - 14 | 1626 | 137 | |
AK220528.1 | - | 4 - 5 | 453 | 137 | ||
AY167030.1 | - | 12 - 13 | 1350 | 137 | ||
BC029195.1 | - | 2 - 3 | 215 | 137 | ||
BC043712.1 | - | 3 - 4 | 353 | 137 | ||
BC063068.1 | - | 2948 | 137 | |||
BC086650.1 | - | 13 - 14 | 1555 | 6849008 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |