Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142701.1 | NP_001136173.1 | 9 - 10 | 1258 | 62 | |
NM_027521.3 | NP_081797.1 | 8 - 9 | 810 | 62 | ||
GenBank mRNA | AK044860.1 | - | 8 - 9 | 772 | 62 | |
AK134029.1 | - | 2 - 3 | 232 | 62 | ||
AK151423.1 | - | 9 - 10 | 1202 | 62 | ||
AK159194.1 | - | 9 - 10 | 1259 | 62 | ||
AK159474.1 | - | 8 - 9 | 1302 | 62 | ||
AK170262.1 | - | 9 - 10 | 1244 | 62 | ||
AK171304.1 | - | 9 - 10 | 1244 | 62 | ||
BC053750.1 | - | 8 - 9 | 810 | 6333971 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |