Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115130.1 | NP_001108602.1 | 6 | 108 | ||
NM_172765.3 | NP_766353.2 | 6 | 108 | |||
GenBank mRNA | AK153955.1 | - | 1 | 108 | ||
BC038156.1 | - | 1 | 108 | |||
BC090965.1 | - | 1 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |