Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164185.1 | NP_001157657.1 | 3 - 4 | 859 | 56 | |
GenBank mRNA | AK167463.1 | - | 3 - 4 | 798 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |