Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028001.3 | NP_082277.2 | 2 - 3 | 154 | 84 | |
GenBank mRNA | BC104136.1 | - | 1 - 2 | 68 | 84 | |
BC104137.1 | - | 1 - 2 | 69 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |