Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291442.1 | NP_001278371.1 | 7 - 8 | 880 | 61 | |
NM_001291443.1 | NP_001278372.1 | 8 - 9 | 874 | 61 | ||
NM_144822.3 | NP_659071.1 | 7 - 8 | 862 | 61 | ||
GenBank mRNA | AK046049.1 | - | 7 - 8 | 845 | 61 | |
AK049521.1 | - | 7 - 8 | 858 | 61 | ||
AK050113.1 | - | 7 - 8 | 849 | 61 | ||
AK052485.1 | - | 7 - 8 | 810 | 61 | ||
AK077605.1 | - | 7 - 8 | 812 | 61 | ||
AK085553.1 | - | 7 - 8 | 781 | 61 | ||
AK200391.1 | - | 4 - 5 | 285 | 61 | ||
AK219825.1 | - | 3 - 4 | 232 | 61 | ||
BC018320.1 | - | 7 - 8 | 803 | 4192966 | 61 | |
BC023022.1 | - | 8 - 9 | 809 | 5367511 | 61 | |
BC023042.1 | - | 6 - 7 | 746 | 5357794 | 61 | |
BC026566.1 | - | 7 - 8 | 813 | 4977083 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |