Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289916.1 | NP_001276845.1 | 5 - 6 | 953 | 78 | |
NM_013646.2 | NP_038674.1 | 6 - 7 | 962 | 78 | ||
GenBank mRNA | AK043990.1 | - | 2 - 3 | 322 | 78 | |
AK149294.1 | - | 5 - 6 | 843 | 78 | ||
AK155827.1 | - | 5 - 6 | 904 | 78 | ||
BC003757.1 | - | 5 - 6 | 902 | 3592667 | 78 | |
U53228.1 | - | 6 - 7 | 946 | 78 | ||
Y08640.1 | - | 5 - 6 | 1108 | 78 | ||
Z82994.1 | - | 5 - 6 | 941 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons |