Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013661.2 | NP_038689.2 | 15 - 16 | 2397 | 102 | |
GenBank mRNA | AK078659.1 | - | 3 - 4 | 329 | 102 | |
AK129362.1 | - | 5 - 6 | 724 | 102 | ||
X97818.1 | - | 15 - 16 | 2188 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |