Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013852.2 | NP_038880.1 | 6 - 7 | 637 | 77 | |
GenBank mRNA | AK030626.1 | - | 6 - 7 | 601 | 77 | |
AK183731.1 | - | 4 - 5 | 339 | 77 | ||
AK189060.1 | - | 4 - 5 | 338 | 77 | ||
AK214630.1 | - | 3 - 4 | 268 | 77 | ||
BC032923.1 | - | 6 - 7 | 578 | 1430389 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |