Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Robo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175549.4 | NP_780758.3 | 26 - 27 | 4324 | 121 | |
GenBank mRNA | AK129396.1 | - | 12 - 13 | 2177 | 121 | |
DQ533875.1 | - | 28 - 29 | 4616 | 121 | ||
DQ533876.1 | - | 26 - 27 | 4324 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |