Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igdcc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290315.1 | NP_001277244.1 | 17 - 18 | 3001 | 58 | |
NM_020043.3 | NP_064427.2 | 17 - 18 | 2998 | 58 | ||
GenBank mRNA | AB052620.1 | - | 17 - 18 | 2998 | 58 | |
AB052621.1 | - | 17 - 18 | 3001 | 58 | ||
AK122535.1 | - | 3077 | 58 | |||
CB524810.1 | - | 4 - 5 | 460 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |