Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033158.2 | NP_001028330.2 | 1 - 2 | 183 | 114 | |
NM_001162923.1 | NP_001156395.1 | 1 - 2 | 183 | 114 | ||
GenBank mRNA | AK014511.1 | - | 1 - 2 | 146 | 113 | |
BC125036.1 | - | 1 - 2 | 160 | 40102054 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |