Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102455.1 | NP_001095925.1 | 11 - 12 | 1674 | 144 | |
NM_001102456.1 | NP_001095926.1 | 10 - 11 | 1506 | 144 | ||
NM_009691.2 | NP_033821.1 | 10 - 11 | 1506 | 144 | ||
GenBank mRNA | AK149977.1 | - | 11 - 12 | 1675 | 144 | |
AK150020.1 | - | 11 - 12 | 1675 | 144 | ||
AK150478.1 | - | 3 - 4 | 406 | 144 | ||
AK151708.1 | - | 3 - 4 | 406 | 144 | ||
AK152787.1 | - | 3 - 4 | 406 | 144 | ||
AK153087.1 | - | 3 - 4 | 406 | 144 | ||
AK164012.1 | - | 11 - 12 | 1692 | 144 | ||
BC051999.1 | - | 10 - 11 | 1484 | 5716148 | 144 | |
BC052396.1 | - | 10 - 11 | 1478 | 5719918 | 144 | |
M97216.1 | - | 6 - 7 | 866 | 144 | ||
U15571.1 | - | 11 - 12 | 1650 | 144 | ||
Z22592.1 | - | 10 - 11 | 1506 | 144 | ||
Z27070.1 | - | 10 - 11 | 1504 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |