Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900002105.mm.1 |
NM_001080817.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002105.mm.1 |
NM_001080817.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002105.mm.1 |
TC0900000279.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900002100.mm.1 |
NM_001080817.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002100.mm.1 |
NM_001080817.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002100.mm.1 |
TC0900000279.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900002101.mm.1 |
NM_001080817.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002101.mm.1 |
NM_001080817.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002101.mm.1 |
TC0900000279.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900002102.mm.1 |
NM_001080817.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002102.mm.1 |
NM_001080817.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002102.mm.1 |
TC0900000279.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900002104.mm.1 |
NM_001080817.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002104.mm.1 |
NM_001080817.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900002104.mm.1 |
TC0900000279.mm.1 |
MTA TC |