Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081152.1 | NP_001074621.1 | 2 - 3 | 255 | 166 | |
GenBank mRNA | AK033012.1 | - | 2 - 3 | 235 | 166 | |
AK080294.1 | - | 2 - 3 | 256 | 166 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |