Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000004958.mm.1 |
NM_029781.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004958.mm.1 |
NM_029781.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004958.mm.1 |
TC1000000707.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000004969.mm.1 |
NM_029781.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004969.mm.1 |
NM_029781.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004969.mm.1 |
TC1000000707.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000004959.mm.1 |
NM_029781.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004959.mm.1 |
NM_029781.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004959.mm.1 |
TC1000000707.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000004957.mm.1 |
NM_029781.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004957.mm.1 |
NM_029781.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004957.mm.1 |
TC1000000707.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000004956.mm.1 |
NM_029781.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004956.mm.1 |
NM_029781.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000004956.mm.1 |
TC1000000707.mm.1 |
MTA TC |