Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027030.2 | NP_081306.1 | 1 - 2 | 222 | 77 | |
GenBank mRNA | AK005584.1 | - | 1 - 2 | 223 | 77 | |
AK078253.1 | - | 1 - 2 | 251 | 77 | ||
AK169641.1 | - | 1 - 2 | 231 | 77 | ||
AK171018.1 | - | 1 - 2 | 214 | 77 | ||
AY040775.1 | - | 1 - 2 | 235 | 77 | ||
BC016273.1 | - | 1 - 2 | 242 | 4511427 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |