Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019727.2 | NP_062701.2 | 2 - 3 | 398 | 61 | |
GenBank mRNA | AB019214.1 | - | 2 - 3 | 338 | 61 | |
AF154120.1 | - | 2 - 3 | 399 | 61 | ||
AK149850.1 | - | 2 - 3 | 310 | 61 | ||
AK154079.1 | - | 2 - 3 | 308 | 61 | ||
AK155746.1 | - | 2 - 3 | 286 | 61 | ||
AK161241.1 | - | 2 - 3 | 379 | 61 | ||
AK167177.1 | - | 2 - 3 | 353 | 61 | ||
AK167993.1 | - | 2 - 3 | 290 | 61 | ||
AK168742.1 | - | 2 - 3 | 391 | 61 | ||
BC066044.1 | - | 2 - 3 | 355 | 6807363 | 61 | |
BC066189.1 | - | 2 - 3 | 289 | 30471747 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |