Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207238.3 | NP_997121.1 | 2 - 3 | 502 | 58 | |
GenBank mRNA | AK053292.1 | - | 2 - 3 | 504 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |