Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198962.3 | NP_945200.1 | 1 - 2 | 294 | 69 | |
GenBank mRNA | AK038551.1 | - | 1 - 2 | 295 | 69 | |
AK048781.1 | - | 1 - 2 | 294 | 69 | ||
AK079572.1 | - | 1 - 2 | 327 | 69 | ||
AY336084.1 | - | 1 - 2 | 262 | 69 | ||
AY336085.1 | - | 1 - 2 | 262 | 69 | ||
BC140957.1 | - | 1 - 2 | 290 | 9055569 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus 1700 Probe spans exons |