Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198962.3 | NP_945200.1 | 3 - 4 | 720 | 86 | |
GenBank mRNA | AF394597.1 | - | 2 - 3 | 349 | 86 | |
AK038551.1 | - | 3 - 4 | 721 | 86 | ||
AK048781.1 | - | 3 - 4 | 720 | 86 | ||
AK079572.1 | - | 3 - 4 | 753 | 86 | ||
AY336084.1 | - | 3 - 4 | 688 | 86 | ||
AY336085.1 | - | 3 - 4 | 688 | 86 | ||
BC140957.1 | - | 3 - 4 | 716 | 9055569 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |