Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164790.1 | NP_001158262.1 | 4 - 5 | 735 | 92 | |
NM_009130.3 | NP_033156.1 | 4 - 5 | 735 | 92 | ||
GenBank mRNA | AK133655.1 | - | 4 - 5 | 637 | 92 | |
AK139140.1 | - | 4 - 5 | 675 | 92 | ||
AK148498.1 | - | 4 - 5 | 717 | 92 | ||
AK158315.1 | - | 4 - 5 | 608 | 92 | ||
AK164089.1 | - | 4 - 5 | 678 | 92 | ||
BC024785.1 | - | 4 - 5 | 694 | 5362975 | 92 | |
U02982.1 | - | 4 - 5 | 699 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |