Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000001649.mm.1 |
NM_001013766.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001649.mm.1 |
NM_001013766.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001649.mm.1 |
NM_001115154.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001649.mm.1 |
NM_001115154.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001649.mm.1 |
TC1000000231.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000001651.mm.1 |
NM_001013766.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001651.mm.1 |
NM_001013766.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001651.mm.1 |
NM_001115154.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001651.mm.1 |
NM_001115154.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001651.mm.1 |
TC1000000231.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000001648.mm.1 |
NM_001013766.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001648.mm.1 |
NM_001013766.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001648.mm.1 |
NM_001115154.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001648.mm.1 |
NM_001115154.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001648.mm.1 |
TC1000000231.mm.1 |
MTA TC |