Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146016.2 | NP_666128.2 | 32 - 33 | 5153 | 92 | |
GenBank mRNA | BC024726.1 | - | 4 - 5 | 588 | 5362808 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |