Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270435.1 | NP_001257364.1 | 12 - 13 | 1986 | 69 | |
NM_001270446.1 | NP_001257375.1 | 7 - 8 | 1242 | 69 | ||
NM_028506.1 | NP_082782.1 | 8 - 9 | 1249 | 69 | ||
NM_029286.2 | NP_083562.1 | 12 - 13 | 2001 | 69 | ||
GenBank mRNA | AK049010.1 | - | 7 - 8 | 1142 | 69 | |
AK078362.1 | - | 12 - 13 | 1928 | 69 | ||
AK163185.1 | - | 12 - 13 | 1927 | 69 | ||
AK224940.1 | - | 8 - 9 | 1178 | 69 | ||
AK224961.1 | - | 7 - 8 | 1241 | 69 | ||
BC114436.1 | - | 12 - 13 | 1928 | 40059415 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |