Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009260.2 | NP_033286.2 | 27 - 28 | 6344 | 61 | |
NM_175836.2 | NP_787030.2 | 27 - 28 | 6004 | 61 | ||
GenBank mRNA | AF017112.1 | - | 27 - 28 | 6251 | 61 | |
AK051630.1 | - | 2 - 3 | 362 | 61 | ||
AK161119.1 | - | 6 - 7 | 1127 | 61 | ||
AK220456.1 | - | 28 - 29 | 6206 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |