Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290753.1 | NP_001277682.1 | 5 - 6 | 1486 | 74 | |
NM_010142.3 | NP_034272.1 | 5 - 6 | 1486 | 74 | ||
GenBank mRNA | BC043088.1 | - | 5 - 6 | 1410 | 6418088 | 74 |
BC062924.1 | - | 5 - 6 | 1431 | 74 | ||
L25890.1 | - | 6 - 7 | 1328 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |