Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133872.2 | NP_598633.2 | 5 - 6 | 957 | 99 | |
GenBank mRNA | AK129170.1 | - | 5 - 6 | 897 | 99 | |
BC059885.1 | - | 5 - 6 | 944 | 6815975 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |