Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pafah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285872.1 | NP_001272801.1 | 3 - 4 | 399 | 93 | |
NM_001285874.1 | NP_001272803.1 | 2 - 3 | 341 | 93 | ||
NM_133880.3 | NP_598641.2 | 3 - 4 | 388 | 93 | ||
GenBank mRNA | AK144090.1 | - | 3 - 4 | 351 | 93 | |
AK156774.1 | - | 2 - 3 | 342 | 93 | ||
BC021890.1 | - | 2 - 3 | 293 | 3598000 | 93 | |
BC025495.1 | - | 3 - 4 | 410 | 5310200 | 93 | |
BC025938.1 | - | 2 - 3 | 234 | 93 | ||
BQ179290.1 | - | 1 - 2 | 110 | 93 | ||
BY032695.1 | - | 3 - 4 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |