Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027402.3 | NP_081678.1 | 1 - 2 | 109 | 69 | |
GenBank mRNA | AJ401376.1 | - | 1 - 2 | 109 | 69 | |
AK005096.1 | - | 1 - 2 | 171 | 69 | ||
BC109183.1 | - | 1 - 2 | 105 | 40050064 | 69 | |
BC109184.1 | - | 1 - 2 | 109 | 40050065 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |