Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017383.3 | NP_059079.2 | 15 - 16 | 2260 | 85 | |
GenBank mRNA | AB032602.1 | - | 14 - 15 | 1946 | 85 | |
AK052972.1 | - | 16 - 17 | 2153 | 85 | ||
BC076594.1 | - | 15 - 16 | 2260 | 30649661 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |