Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170488.1 | NP_001163959.1 | 5 - 6 | 557 | 171 | |
NM_176842.3 | NP_789812.1 | 4 - 5 | 449 | 171 | ||
GenBank mRNA | BC024858.1 | - | 4 - 5 | 411 | 5357332 | 171 |
BC027162.1 | - | 5 - 6 | 538 | 4951567 | 171 | |
BC027413.1 | - | 4 - 5 | 436 | 4022441 | 171 | |
BM948212.1 | - | 2 - 3 | 186 | 171 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |