Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011708.4 | NP_035838.3 | 11 - 12 | 1546 | 71 | |
GenBank mRNA | AF539800.2 | - | 10 - 11 | 1294 | 71 | |
AK044921.1 | - | 11 - 12 | 1533 | 71 | ||
AK157296.1 | - | 11 - 12 | 1548 | 71 | ||
AY208897.1 | - | 11 - 12 | 1354 | 71 | ||
DQ355287.1 | - | 11 - 12 | 1531 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |