Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013528.3 | NP_038556.1 | 14 - 15 | 1582 | 127 | |
GenBank mRNA | AF334736.1 | - | 15 - 16 | 1587 | 127 | |
AK019852.1 | - | 14 - 15 | 1578 | 127 | ||
BC002283.1 | - | 11 - 12 | 1181 | 127 | ||
BC010516.1 | - | 14 - 15 | 1530 | 4191553 | 127 | |
BC050762.1 | - | 14 - 15 | 1578 | 6742987 | 127 | |
U00932.1 | - | 13 - 14 | 1429 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |