Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033253.3 | NP_001028425.1 | 14 - 15 | 1621 | 112 | |
NM_001159942.1 | NP_001153414.1 | 15 - 16 | 1787 | 112 | ||
GenBank mRNA | AK041942.1 | - | 3 - 4 | 114 | 112 | |
AK122468.1 | - | 11 - 12 | 945 | 112 | ||
AK135923.1 | - | 14 - 15 | 1623 | 112 | ||
BC076628.1 | - | 12 - 13 | 1018 | 112 | ||
BC117965.1 | - | 13 - 14 | 1223 | 40098326 | 112 | |
BC117966.1 | - | 13 - 14 | 1216 | 40098330 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |