Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021899.3 | NP_068699.1 | 2 - 3 | 1133 | 74 | |
GenBank mRNA | AB221666.1 | - | 1 - 2 | 332 | 74 | |
AF253052.1 | - | 2 - 3 | 1145 | 74 | ||
AK154574.1 | - | 2 - 3 | 1376 | 74 | ||
BC040395.1 | - | 3 - 4 | 763 | 4190087 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |