Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175628.3 | NP_783327.2 | 30 - 31 | 4091 | 84 | |
GenBank mRNA | AY185125.1 | - | 30 - 31 | 4037 | 84 | |
BC023844.1 | - | 20 - 21 | 2753 | 84 | ||
BC072642.1 | - | 30 - 31 | 4031 | 6839761 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |