Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600022734.mm.1 |
TC0600002958.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022734.mm.1 |
NM_001168256.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022734.mm.1 |
NM_001168256.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022734.mm.1 |
NM_001168257.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022734.mm.1 |
NM_001168257.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022734.mm.1 |
NM_144805.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022734.mm.1 |
NM_144805.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022735.mm.1 |
TC0600002958.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022735.mm.1 |
NM_001168256.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022735.mm.1 |
NM_001168256.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022735.mm.1 |
NM_001168257.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022735.mm.1 |
NM_001168257.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022735.mm.1 |
NM_144805.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022735.mm.1 |
NM_144805.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022737.mm.1 |
TC0600002958.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022737.mm.1 |
NM_144805.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022737.mm.1 |
NM_144805.2 |
Refseq |