Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282126.1 | NP_001269055.1 | 9 - 10 | 3342 | 63 | |
NM_001282127.1 | NP_001269056.1 | 9 - 10 | 3107 | 63 | ||
NM_001282128.1 | NP_001269057.1 | 9 - 10 | 3357 | 63 | ||
NM_030178.2 | NP_084454.1 | 9 - 10 | 3107 | 63 | ||
GenBank mRNA | AK019522.1 | - | 3 - 4 | 734 | 63 | |
AK156375.1 | - | 9 - 10 | 3290 | 63 | ||
BC005647.1 | - | 7 - 8 | 1379 | 63 | ||
BC031594.1 | - | 7 - 8 | 2031 | 63 | ||
BC046521.1 | - | 9 - 10 | 3047 | 5363697 | 63 | |
BC138361.1 | - | 9 - 10 | 2931 | 8861381 | 63 | |
BC145256.1 | - | 9 - 10 | 3181 | 9053809 | 63 | |
DQ288860.1 | - | 4 - 5 | 1797 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |