Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033463.3 | NP_001028635.2 | 2 - 3 | 508 | 90 | |
GenBank mRNA | AK146408.1 | - | 1 - 2 | 257 | 90 | |
AK156197.1 | - | 2 - 3 | 509 | 90 | ||
AK220217.1 | - | 1 - 2 | 439 | 90 | ||
BC141065.1 | - | 2 - 3 | 443 | 9055677 | 90 | |
BC145318.1 | - | 2 - 3 | 443 | 9053873 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |