Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175101.3 | NP_780310.1 | 4 - 5 | 559 | 67 | |
GenBank mRNA | AK075595.1 | - | 4 - 5 | 560 | 67 | |
AK167819.1 | - | 4 - 5 | 408 | 67 | ||
AK210449.1 | - | 4 - 5 | 269 | 67 | ||
BC004641.1 | - | 4 - 5 | 555 | 3157147 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |