Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clstn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153508.4 | NP_705728.1 | 1 - 2 | 364 | 73 | |
GenBank mRNA | AJ278486.1 | - | 1 - 2 | 231 | 73 | |
AK032336.1 | - | 1 - 2 | 306 | 73 | ||
BC055054.1 | - | 1 - 2 | 296 | 6493096 | 73 | |
CJ152135.1 | - | 1 - 2 | 365 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |