Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhdds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026144.4 | NP_080420.2 | 5 - 6 | 593 | 68 | |
GenBank mRNA | AK014317.1 | - | 6 - 7 | 781 | 68 | |
AK034277.1 | - | 5 - 6 | 655 | 68 | ||
AK036929.1 | - | 5 - 6 | 671 | 68 | ||
AK049991.1 | - | 5 - 6 | 594 | 68 | ||
AK135087.1 | - | 5 - 6 | 655 | 68 | ||
AK149138.1 | - | 5 - 6 | 625 | 68 | ||
AK171207.1 | - | 5 - 6 | 667 | 68 | ||
BC004011.1 | - | 5 - 6 | 641 | 3493585 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |