Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scmh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159630.1 | NP_001153102.1 | 8 - 9 | 929 | 135 | |
NM_013883.2 | NP_038911.1 | 8 - 9 | 929 | 135 | ||
GenBank mRNA | AB030906.1 | - | 8 - 9 | 828 | 135 | |
AB221680.1 | - | 4 - 5 | 470 | 135 | ||
AB221681.1 | - | 4 - 5 | 470 | 135 | ||
BC034667.1 | - | 6 - 7 | 639 | 5356322 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |