Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134023.1 | NP_598784.1 | 4 - 5 | 584 | 97 | |
GenBank mRNA | AK140872.1 | - | 4 - 5 | 596 | 97 | |
AK153677.1 | - | 566 | 97 | |||
BC018300.1 | - | 4 - 5 | 584 | 4193046 | 97 | |
BC029773.1 | - | 2 - 3 | 148 | 97 | ||
BC037230.1 | - | 4 - 5 | 567 | 2650923 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |