Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178149.4 | NP_835362.2 | 4 - 5 | 718 | 91 | |
GenBank mRNA | AF528078.1 | - | 4 - 5 | 642 | 91 | |
AF528081.1 | - | 4 - 5 | 643 | 91 | ||
AK154842.1 | - | 4 - 5 | 660 | 91 | ||
AK160300.1 | - | 2 - 3 | 1713 | 91 | ||
AY522649.1 | - | 4 - 5 | 1049 | 91 | ||
BC055920.1 | - | 4 - 5 | 655 | 4039129 | 91 | |
BC079641.1 | - | 4 - 5 | 642 | 5707778 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |