Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252250.1 | NP_001239179.1 | 9 - 10 | 1440 | 123 | |
NM_001252251.1 | NP_001239180.1 | 9 - 10 | 1440 | 123 | ||
NM_001252252.1 | NP_001239181.1 | 9 - 10 | 1440 | 123 | ||
NM_001252253.1 | NP_001239182.1 | 9 - 10 | 905 | 123 | ||
NM_010898.4 | NP_035028.2 | 9 - 10 | 1440 | 123 | ||
GenBank mRNA | AK017343.1 | - | 9 - 10 | 906 | 123 | |
AK045998.1 | - | 9 - 10 | 1262 | 123 | ||
AK167685.1 | - | 6 - 7 | 531 | 123 | ||
BC005442.1 | - | 9 - 10 | 1306 | 3496053 | 123 | |
DV654856.1 | - | 6 - 7 | 498 | 123 | ||
L22987.1 | - | 9 - 10 | 898 | 123 | ||
L22988.1 | - | 9 - 10 | 898 | 123 | ||
L22989.1 | - | 9 - 10 | 898 | 123 | ||
L27090.1 | - | 9 - 10 | 891 | 123 | ||
L27105.1 | - | 9 - 10 | 891 | 123 | ||
L28176.1 | - | 9 - 10 | 1198 | 123 | ||
X74671.1 | - | 9 - 10 | 1457 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |